Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Trichostatin A, Histone Deacetylase Inhibitor, >98% Purity, Solid, 1 mg

Catalog No. 502707197
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-7197 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-7197 Supplier Abcam Supplier No. AB1208501MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Potent and selective histone deacetylase (HDAC) inhibitor with anticancer and antifungal activity.

  • Potency: Potent, selective histone deacetylase (HDAC) inhibitor (Ki = 3.4 nM)
  • Purity: >98%
  • Molecular Weight: 302.37 Da
  • Form: Solid
  • Applications: Induces cell proliferation, differentiation and apoptosis in vitro; anticancer and antifungal agent; active in vivo and in vitro
  • Storage: Store at -20°C

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Buffer 100% Trichostatin A
Content And Storage Store at -20°C.
Form Solid
Shipping Condition Ambient
Substrate Properties Chemical Substrate
Target Molecule Histone Deacetylase
Formulation 100% Trichostatin A
Quantity 1 mg
For Use With (Application) Cancer research, Antifungal research, Cell proliferation studies, Cell differentiation studies, Apoptosis studies
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.