Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Trichostatin A, Histone Deacetylase Inhibitor, Liquid, 1 mg

Catalog No. 502704557
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-4557 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-4557 Supplier Abcam Supplier No. AB1465981MGLQD
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potent and selective histone deacetylase inhibitor with >99% purity.

  • Potency: Potent and selective histone deacetylase (HDAC) inhibitor with Ki = 3.4 nM
  • Purity: >99%
  • Molecular Weight: 302.37 Da
  • Biological Activity: Induces cell proliferation, differentiation, and apoptosis in vitro; possesses anticancer and antifungal properties
  • Form: Liquid
  • Storage Buffer: 99.73% Dimethylsulfoxide, 0.27% Trichostatin A

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Content And Storage Store at -20°C.
Form Liquid
Buffer 99.73% dimethylsulfoxide, 0.27% Trichostatin A
Shipping Condition Ambient
Substrate Properties Chemical Substrate
Target Molecule Histone Deacetylase
Formulation 99.73% Dimethylsulfoxide, 0.27% Trichostatin A
Quantity 1 mg
For Use With (Application) HDAC inhibition, cell proliferation, cell differentiation, apoptosis, cancer research, antifungal research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.