Learn More
StatLab TRICHROME STAIN KIT MASSON'S
SDP
Supplier:  StatLab KTMTR2
Encompass_Preferred
Bouin's 2000™ 100ml, Biebrich Scarlet/Acid Fucshin 100ml, Phosphomolybdic/Phosphotungstic Acid 100ml, Aniline Blue Stain 100ml, 1% Acetic Acid 100ml and Weigert's A & B Hematoxylin 100ml each
Catalog No. NC9691313
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.