Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tricyclo[5.2.1.0(2,6)]decanedimethanol 90.0+%, TCI America™
SDP

Catalog No. T085025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T085025G 25 g
T0850500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T085025G Supplier TCI America Supplier No. T085025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 26896-48-0
Molecular Formula C12H20O2
MDL Number MFCD00151166
Synonym Bis(hydroxymethyl)tricyclo[5.2.1.0(2,6)]decane, Tricyclodecanedimethanol (so called)

Specifications

Color Colorless
Boiling Point 175°C
Quantity 25 g
Formula Weight 196.29
Percent Purity ≥90.0% (GC)
Physical Form Starch Syrup
Chemical Name or Material Tricyclo[5.2.1.0(2,6)]decanedimethanol
EINECSNumber (4)-0641
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.