Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trielaidin 96.0+%, TCI America™
SDP

Catalog No. T13925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T13925G 5 g
T13921G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T13925G Supplier TCI America Supplier No. T13925G

Chemical Identifiers

CAS 537-39-3
Molecular Formula C57H104O6
Molecular Weight (g/mol) 885.45
MDL Number MFCD00067496
InChI Key PHYFQTYBJUILEZ-WUOFIQDXSA-N
Synonym Glycerol Elaidate, 1,2,3-Tri(trans-9-octadecenoyl) Glycerol
PubChem CID 5364673
ChEBI CHEBI:53759
IUPAC Name 1,3-bis[(9E)-octadec-9-enoyloxy]propan-2-yl (9E)-octadec-9-enoate
SMILES CCCCCCCC\C=C\CCCCCCCC(=O)OCC(COC(=O)CCCCCCC\C=C\CCCCCCCC)OC(=O)CCCCCCC\C=C\CCCCCCCC

Specifications

Melting Point 42°C
Color White-Yellow
Quantity 5 g
Formula Weight 885.45
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Trielaidin
EINECSNumber (2)-0669
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.