Learn More
Research Products International Corp Triethanolamine, 4 L
SDP
Supplier:  Research Products International Corp T230404.0
Encompass_Preferred
Common Name: 2,2',2"-NitrilothriethanolCAS Number: 102-71-6Molecular Weight: 149.2Chemical Formula: C6H15NO3Solubility: WaterSTOR Temperature: Room TemperatureSTOR Conditions: Room TemperatureAppearance: Clear to light liquid Assay: 99.0 %Color, PTCO: 40 Diethanolamine : 0.50 %Monoethanolamine: 0.10 %Water (KF): 0.5 %Shipping Weight: 1.90 ozShipping Dimensions: 6.75 x 11.50 x 3.50Product usually ships within 24 hours.Research or further manufacturing use only, not for food or drug use.
Catalog No. 50-223-0303
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.