Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethanolamine, 97 to 98%, MP Biomedicals™

Catalog No. ICN15216391 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15216391 1 L
ICN15216380 100 mL
MP215216390 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15216391 Supplier MP Biomedicals, Inc Supplier No. 0215216391
Add to Cart
Add to Cart

Triethanolamine ≥97%, Liquid

  • It is used as an emulsifier and a surfactant. It finds application in common products like dishwashing liquids, general cleaners, hand cleaners, shaving cream, metalworking fluids, liquid laundry detergents, paints and printing inks.
  • It is used for the neutralization of fatty acids and adjusting buffer pH as well as utilized to dissolve oils. It acts as an effective complexing agent in electroless plating.
  • It is used in hair care products like hair dyes and wave sets.
  • Protein purification reagent Triethanolamine acts as an organic additive in the grinding of cement clinker and as a complexing agent for aluminum ions in aqueous solutions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9005-65-6

Specifications

Color Colorless to Yellow
Density 1.124g/cm3 at 20°C
Assay Percent Range 97 to 98%
Molecular Formula (C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6
MDL Number MFCD00082107
Quantity 1 L
Synonym Daltogen, Nitrilo-2, 2, 2-Triethanol
InChI Key HDTIFOGXOGLRCB-KTKRTIGZNA-N
SMILES CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
IUPAC Name 2-{2-[3,5-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl (9E)-octadec-9-enoate
Molecular Weight (g/mol) 604.82
Formula Weight 149.2
Physical Form Liquid
Chemical Name or Material Triethanolamine
Show More Show Less
EINECSNumber 203-049-8
storageNote1 Store Desiccated.. Store Under Nitrogen. Protect from light
Recommended Storage Store at Room Temperature (15° to 30°C)
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.