Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethanolamine, 98+%, Thermo Scientific Chemicals

Catalog No. AAL044860E
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL044860E 2500 g
AAL0448622 100 g
AAL0448636 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL044860E Supplier Thermo Scientific Chemicals Supplier No. L044860E
Add to Cart

Protein purification reagent Triethanolamine acts as an organic additive in the grinding of cement clinker and as a complexing agent for aluminum ions in aqueous solutions. It is used as an emulsifier and a surfactant. It finds application in common products like dishwashing liquids, general cleaners, hand cleaners, shaving cream, metalworking fluids, liquid laundry detergents, paints and printing inks. It is used for the neutralization of fatty acids and adjusting buffer pH as well as utilized to dissolve oils. It acts as an effective complexing agent in electroless plating. Further, it is used in hair care products like hair dyes and wave sets.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Protein purification reagent Triethanolamine acts as an organic additive in the grinding of cement clinker and as a complexing agent for aluminum ions in aqueous solutions. It is used as an emulsifier and a surfactant. It finds application in common products like dishwashing liquids, general cleaners, hand cleaners, shaving cream, metalworking fluids, liquid laundry detergents, paints and printing inks. It is used for the neutralization of fatty acids and adjusting buffer pH as well as utilized to dissolve oils. It acts as an effective complexing agent in electroless plating. Further, it is used in hair care products like hair dyes and wave sets.

Solubility
Miscible with water, methanol, acetone, ether, carbon tetrachloride, n-heptane and chloroform. Slightly soluble in petroleum ether.

Notes
Air sensitive and hygroscopic Incompatible with acids, oxidizing agents, reducing agents and metals.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102-71-6
Molecular Formula C6H15NO3
Molecular Weight (g/mol) 149.19
MDL Number MFCD00002855
InChI Key GSEJCLTVZPLZKY-UHFFFAOYSA-N
Synonym triethanolamine, trolamine, 2,2',2-nitrilotriethanol, sterolamide, daltogen, tris 2-hydroxyethyl amine, triethylolamine, nitrilotriethanol, trihydroxytriethylamine, thiofaco t-35
PubChem CID 7618
ChEBI CHEBI:28621
IUPAC Name 2-[bis(2-hydroxyethyl)amino]ethan-1-ol
SMILES OCCN(CCO)CCO

Specifications

Melting Point 19°C to 21°C
Density 1.124
Boiling Point 206°C to 207°C (15 mmHg)
Flash Point 185°C (365°F)
Odor Ammonia-like
Refractive Index 1.485
Linear Formula (HOCH2CH2)3N
Quantity 2500 g
Beilstein 1699263
Sensitivity Hygroscopic; Light sensitive
Merck Index 14,9665
Solubility Information Miscible with water,methanol,acetone,ether,carbon tetrachloride,n-heptane and chloroform. Slightly soluble in petroleum ether.
Formula Weight 149.19
Percent Purity ≥98%
Chemical Name or Material Triethanolamine
Show More Show Less
Hazard Category H317-H373
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P260-P280-P302+P352-P314-P333+P313-P362-P501c
EINECSNumber 203-049-8
RTECSNumber KL9275000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.