Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethoxy-n-octylsilane 97.0+%, TCI America™
SDP

Catalog No. O0171100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O0171100ML 100 mL
O017125ML 25 mL
O0171500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. O0171100ML Supplier TCI America Supplier No. O0171100ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2943-75-1
Molecular Formula C14H32O3Si
Molecular Weight (g/mol) 276.49
MDL Number MFCD00039883
InChI Key MSRJTTSHWYDFIU-UHFFFAOYSA-N
Synonym n-octyltriethoxysilane, octyltriethoxysilane, triethoxy octyl silane, silane, triethoxyoctyl, dynasylan octeo, octyl triethoxy silane, prosil 9202, silquest a 137, prosil 9234, a 137 coupling agent
PubChem CID 76262
IUPAC Name triethoxy(octyl)silane
SMILES CCCCCCCC[Si](OCC)(OCC)OCC

Specifications

Color Colorless
Boiling Point 106°C
Quantity 100 mL
Formula Weight 276.49
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Triethoxy-n-octylsilane
EINECSNumber (2)-3784
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.