Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethyl 3-Phosphonopropionate 98.0+%, TCI America™
SDP

Catalog No. D1524250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1524250G 250 g
D152425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D1524250G Supplier TCI America Supplier No. D1524250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3699-67-0
Molecular Formula C9H19O5P
Molecular Weight (g/mol) 238.22
MDL Number MFCD00015167
InChI Key JMJWCUOIOKBVNQ-UHFFFAOYSA-N
Synonym ethyl 3-diethoxyphosphoryl propanoate, triethyl 3-phosphonopropionate, 3-diethylphosphonopropionic acid ethyl ester, triethyl 3-phosphonopropanoate, ethyl 3-diethoxyphosphinyl propionate, ethyl 3-diethoxycarbonyl propanoate, propanoic acid, 3-diethoxyphosphinyl-, ethyl ester, ethyl 3-diethylphosphonopropionate, ethyl 3-diethylphosphono-propionate, ethyl 3-diethylphosphono propanoate
PubChem CID 281204
IUPAC Name ethyl 3-diethoxyphosphorylpropanoate
SMILES CCOC(=O)CCP(=O)(OCC)OCC

Specifications

Color Colorless
Boiling Point 95°C
Quantity 250 g
Formula Weight 238.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Triethyl 3-Phosphonopropionate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.