Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethyl Orthochloroacetate 95.0+%, TCI America™
SDP

Catalog No. C12881ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C12881ML 1 mL
C12885ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C12881ML Supplier TCI America Supplier No. C12881ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 51076-95-0
Molecular Formula C8H17ClO3
Molecular Weight (g/mol) 196.671
MDL Number MFCD00142889
InChI Key URFKLQSFBXBOQU-UHFFFAOYSA-N
Synonym 2-chloro-1,1,1-triethoxy-ethane, triethyl orthochloroacetate, 2-chloro-1,1,1-triethoxy ethane, orthochloroacetic acid triethyl ester, ethane, 2-chloro-1,1,1-triethoxy, chlorotriethoxyethane, acmc-209tex, 2-chloro-1, 1,1-triethoxy-ethane, 2-chloranyl-1,1,1-triethoxy-ethane, 2-chloro-1,1,1-triethoxyethane
PubChem CID 305922
IUPAC Name 2-chloro-1,1,1-triethoxyethane
SMILES CCOC(CCl)(OCC)OCC

Specifications

Color Colorless
Boiling Point 91°C
Quantity 1 mL
Formula Weight 196.67
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Triethyl Orthochloroacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.