Learn More
Sigma Organic Chemistry Triethylamine | 1L | 121-44-8 | MFCD00009051
Supplier: Sigma Organic Chemistry T08861L

Triethylamine, 1L
About This Item:
Synonym(s): TEA; N,N-Diethylethanamine
Linear Formula: (C2H5)3N
Molecular Weight: 101.19
CAS Number: 121-44-8
MDL Number: MFCD00009051
UNSPSC Code: 12352116
Purity: >=99%
Boiling Point: 88.8 C (lit.)
Melting Point: -115 C (lit.)
Density: 0.726 g/mL at 25 C (lit.)
Refractive Index: n20/D 1.401 (lit.)
Storage: room temp
EINECS Number: 204-469-4
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.