Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethylamine, BAKER BIO-ANALYZED™ Reagent, J.T. Baker™

Catalog No. 02004537 Shop All Avantor Products
Change view
Click to view available options
Quantity:
12 x 500 mL
Packaging:
Narrow Mouth Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02-004-537 12 x 500 mL Narrow Mouth Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-004-537 Supplier Avantor Supplier No. 911107
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specially purified to assure suitability in nucleic acid or peptide synthesis

Chemical Identifiers

CAS 121-44-8
Molecular Formula C6H15N
Molecular Weight (g/mol) 101.193
InChI Key ZMANZCXQSJIPKH-UHFFFAOYSA-N
Synonym triethylamine, ethanamine, n,n-diethyl, diethylamino ethane, triaethylamin, triethylamin, trietilamina, triethyl amine, n,n,n-triethylamine, net3, diethylaminoethane
PubChem CID 8471
ChEBI CHEBI:35026
IUPAC Name N,N-diethylethanamine
SMILES CCN(CC)CC

Specifications

Flash Point −11.00°C
Packaging Narrow Mouth Amber Glass Bottle
Quantity 12 x 500 mL
Specific Gravity 1L = 0.73kg
Formula Weight 101.19
Percent Purity 100%
Grade BAKER BIO-ANALYZED™ Reagent
Physical Form Liquid
Chemical Name or Material Triethylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.