Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethylamine, BioSyn™, for biosynthesis, for HPLC, >99.5%, Solstice

Catalog No. 60048599 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
12 x 100 mL
6 x 1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-048-599 12 x 100 mL
60-048-598 6 x 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-048-599 Supplier Solstice Supplier No. BB359P100

missing translation for 'validMainframeMsg'

  • Ultra-low water specifications
  • Low amine specifications
  • Low particulate levels
  • High solvent purity
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-44-8
Molecular Formula C6H15N
Molecular Weight (g/mol) 101.19
InChI Key ZMANZCXQSJIPKH-UHFFFAOYSA-N
IUPAC Name triethylamine
SMILES CCN(CC)CC

Specifications

CAS Max % 100.00
Density 0.729 g/cm3
Boiling Point 89.5°C
Flash Point -7°C
Packaging Glass Bottle
Quantity 12 x 100 mL
Grade HPLC
Physical Form Liquid
Hazard Category Danger
Hazard Statement H225+H302+H311+H331+H314+H335
Precautionary Statement P280+P271+P270+P264+P261+P243+P242+P241+P240+P233+P210
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.