Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethylamine, MilliporeSigma™

Catalog No. MTX12005 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MTX12005 500 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MTX12005 Supplier MilliporeSigma Supplier No. TX12005
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Solvent designed for routine chemical synthesis, laboratory applications, and quality control environments.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-44-8
Molecular Formula C6H15N
Molecular Weight (g/mol) 101.193
InChI Key ZMANZCXQSJIPKH-UHFFFAOYSA-N
Synonym triethylamine, ethanamine, n,n-diethyl, diethylamino ethane, triaethylamin, triethylamin, trietilamina, triethyl amine, n,n,n-triethylamine, net3, diethylaminoethane
PubChem CID 8471
ChEBI CHEBI:35026
IUPAC Name N,N-diethylethanamine
SMILES CCN(CC)CC

Specifications

Density 0.73g/cm3 (20°C)
Explosion Limit 1.2 to 9.3%(V)
Ignition Point 215°C
Packaging Glass Bottle
Linear Formula (C2H5)3N
Quantity 500 mL
Vapor Pressure 69 hPa (20°C)
Formula Weight 11.19
Percent Purity ≥99.5%
Grade Not Available
Physical Form Liquid
Chemical Name or Material Triethylamine, For Synthesis
Show More Show Less
Recommended Storage Store at +15°C to +25°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.