Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethylamine, Solstice

Catalog No. 60090602 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
10 x 2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-090-602 10 x 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 60-090-602 Supplier Solstice Supplier No. 1792410X2ML

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For HPLC, ≥99.5%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-44-8
PubChem CID 8471
ChEBI CHEBI:35026

Specifications

Percent Purity ≥99.5%
Grade HPLC
Packaging Ampoule
Quantity 10 x 2 mL
Flash Point -11 °C
Physical Form Liquid
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.