Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethylamine Hydrochloride, ≥99%, For HPLC, Solstice

Catalog No. 60047782 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
50 g
250 g
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-047-782 250 g Poly Bottle
60-047-783 50 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-047-782 Supplier Solstice Supplier No. 96249250G

Chemical Identifiers

CAS 554-68-7
Molecular Formula C6H16ClN
Molecular Weight (g/mol) 137.65
MDL Number MFCD00012500
InChI Key ILWRPSCZWQJDMK-UHFFFAOYSA-N
Synonym triethylamine hydrochloride, triethylammonium chloride, triethylamine hcl, ethanamine, n,n-diethyl-, hydrochloride, triethylamine monohydrochloride, triethylamine, hydrochloride, unii-0nx3818gcw, amine, triethyl, hydrochloride, n,n-diethylethanamine hydrochloride, triethylaminehydrochloride
PubChem CID 11130
IUPAC Name hydrogen triethylamine chloride
SMILES [H+].[Cl-].CCN(CC)CC

Specifications

Melting Point 261°C
Packaging Poly Bottle
Linear Formula (C2H5)3N · HCl
Quantity 250 g
UN Number NONH for all modes of transport
Beilstein 3906409
Formula Weight 137.65g/mol
Percent Purity ≥99%
Chemical Name or Material Triethylamine hydrochloride
Hazard Statement P261-P305 + P351 + P338
RTECSNumber 209-067-2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.