Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ProteoSpec™ Triethylamine, LCMS Grade, Ricca Chemical
SDP

Catalog No. MB11205101 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
10 x 1 mL
Packaging:
Glass Ampule
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MB11205101 10 x 1 mL Glass Ampule
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MB11205101 Supplier Ricca Chemical Company Supplier No. RMB1120510X1R

missing translation for 'validMainframeMsg'

LCMS Mobile Phase

Chemical Identifiers

CAS 121-44-8
Molecular Formula C6H15N
Molecular Weight (g/mol) 101.193
InChI Key ZMANZCXQSJIPKH-UHFFFAOYSA-N
Synonym triethylamine, ethanamine, n,n-diethyl, diethylamino ethane, triaethylamin, triethylamin, trietilamina, triethyl amine, n,n,n-triethylamine, net3, diethylaminoethane
PubChem CID 8471
ChEBI CHEBI:35026
IUPAC Name N,N-diethylethanamine
SMILES CCN(CC)CC

Specifications

CAS Min % 100
CAS Max % 100
Melting Point -115°C
Color Colorless
Density 0.73
Boiling Point 90°C
Packaging Glass Ampule
Quantity 10 x 1 mL
Formula Weight 101.19
Percent Purity 99.5% min.
Grade LC-MS
Physical Form Liquid
Assay GC
Chemical Name or Material ProteoSpec™ Triethylamine, LCMS Grade
Show More Show Less
ShelfLife 12 months
Recommended Storage Ambient
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.