Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethylamine, Reagent, 99%, Spectrum™ Chemical
SDP

Catalog No. 18607571 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-571 500 mL Amber Glass Bottle
18-607-572 4 L Amber Glass Bottle
18-607-573 20 L Amber Glass Bottle
18-607-574 200 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-607-571 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. TR117500MLGL
Add to Cart
Add to Cart

TR117, 121-44-8, C6H15N

Spectrum™ Chemical Triethylamine, Reagent is a chemical compound that is often referred to as TEA. This volatile liquid has no color and has a pungent fish like smell combined with ammonia. Most often used as a base in organic synthesis. It is also specifically used as the active ingredient of mosquito anesthetic. Its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.

Chemical Identifiers

CAS 121-44-8
Molecular Formula C6H15N
Molecular Weight (g/mol) 101.19
InChI Key ZMANZCXQSJIPKH-UHFFFAOYSA-N
IUPAC Name triethylamine
SMILES CCN(CC)CC

Specifications

CAS Max % 1
Boiling Point 89.7°C / 193.5°F
Packaging Amber Glass Bottle
Quantity 500 mL
Percent Purity 99%
Grade Reagent
Physical Form Viscous Liquid or Low Melting Solid
Chemical Name or Material Triethylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.