Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Solstice Triethylammonium acetate buffer, BioUltra, volatile buffer, ∼1.0 M in H2O, Solstice

Catalog No. 60047739 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-047-739 100 mL
600-28-77 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-047-739 Supplier Solstice Supplier No. 90357100ML
Add to Cart
Add to Cart

BioUltra, volatile buffer, ∼1.0 M in H2O

  • Finest quality products for life science
  • The most stringent testing protocols and highest purity
  • Superior manufacturing methods and product specific testing protocols designed to ensure quality to our highest standard
  • Tests may include, but are not limited to, purity by gel electrophoresis, trace metal testing and application testing.
TRUSTED_SUSTAINABILITY

Specifications

CAS 5204-74-0
Physical Form Solution
Quantity 100 mL
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.