Learn More
Solstice Triethylammonium Bicarbonate Buffer, Solstice
Shop All Solstice Research Chemicals ProductsDescription
- Leading the Industry For Over 65 Years
- Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| CAS | 15715-58-9 |
| Color | Colorless |
| For Use With (Equipment) | Volatile buffer for HPLC |
| pH | 8.4 to 8.6 (at 25°C) |
| Physical Form | Liquid |
| Quantity | 500 mL |
| Packaging | Glass Bottle |
| Molecular Weight (g/mol) | 163.217 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.