Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethylene Glycol Monobutyl Ether 98.0+%, TCI America™
SDP

Catalog No. T091525ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T091525ML 25 mL
T0915500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T091525ML Supplier TCI America Supplier No. T091525ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 143-22-6
Molecular Formula C10H22O4
Molecular Weight (g/mol) 206.28
MDL Number MFCD00020606
InChI Key COBPKKZHLDDMTB-UHFFFAOYSA-N
Synonym triethylene glycol monobutyl ether, butoxytriglycol, dowanol tbat, butoxytriethylene glycol, poly-solv tb, triethylene glycol butyl ether, triglycol monobutyl ether, ethanol, 2-2-2-butoxyethoxy ethoxy, triethylene glycol mono-n-butyl ether, triethylene glycol n-butyl ether
PubChem CID 8923
IUPAC Name 2-[2-(2-butoxyethoxy)ethoxy]ethan-1-ol
SMILES CCCCOCCOCCOCCO

Specifications

Melting Point -35°C
Color Yellow
Boiling Point 278°C
Quantity 25 mL
Formula Weight 206.28
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Triethylene Glycol Monobutyl Ether
EINECSNumber (2)-0436&(7)-0097
RTECSNumber KJ9450000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.