Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethylene Glycol, Purified, 97%, Spectrum™ Chemical
SDP

Catalog No. 18614554 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
200 L
Packaging:
Steel Can
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-614-554 4 L Steel Can
18-614-552 500 mL Steel Can
18-614-551 20 L Steel Can
18-614-553 200 L Steel Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-614-554 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. T11184LTGL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 112-27-6
Molecular Formula C6H14O4
Molecular Weight (g/mol) 150.17
InChI Key ZIBGPFATKBEMQZ-UHFFFAOYSA-N
IUPAC Name 2-[2-(2-hydroxyethoxy)ethoxy]ethan-1-ol
SMILES OCCOCCOCCO

Specifications

CAS Max % 1
Boiling Point 285°C to 287.4°C (545°F to 549.32°F)
Packaging Steel Can
Quantity 4 L
Percent Purity 0.97
Grade Purified
Physical Form Viscous Liquid or Low Melting Solid
Chemical Name or Material Triethylene Glycol

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.