accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOOOH

CAS RN 112-27-6

IUPAC Name: 2-[2-(2-hydroxyethoxy)ethoxy]ethan-1-ol
Synonyms: triethylene glycol caswell no. 888 triglycol trigen TEG 2,2'-ethylenedioxyethanol dipotassium salt triethylene glycol triethylenglykol bis(2-hydroxyethoxyethane) disodium salt triethylene glycol
Molecular Weight (g/mol): 150.17
Molecular Formula: C6H14O4
InChi Key: ZIBGPFATKBEMQZ-UHFFFAOYSA-N
SMILES: OCCOCCOCCO
Molecular Weight (g/mol) 
  • (13)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (3)
  • (1)
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (4)
  • (6)
Physical Form 
  • (3)
  • (3)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (10)
  • (4)
  • (3)
  • (4)
  • (1)
  • (2)
  • (6)
  • (13)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
  • (4)
  • (2)
  • (2)
  • (4)
  • (6)
  • (3)
  • (3)
  • (4)
  • (4)
  • (1)
  • (1)