Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethyloxonium tetrafluoroborate, 95%, stab. with 3-5% diethyl ether

Catalog No. AAA1442022
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1442022 100 g
AAA1442014 25 g
AAA1442036 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1442022 Supplier Thermo Scientific Chemicals Supplier No. A1442022
Add to Cart

CAS: 368-39-8 | C6H15BF4O | 189.99 g/mol

Triethyloxonium tetrafluoroborate is used as a powerful alkylating agent, especially for ethylation. It is used for the modification of carboxyl residues in proteins. It is involved in the preparation of μ -amino esters from lactams.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triethyloxonium tetrafluoroborate is used as a powerful alkylating agent, especially for ethylation. It is used for the modification of carboxyl residues in proteins. It is involved in the preparation of µ -amino esters from lactams.

Solubility
Soluble in dichloromethane.

Notes
Moisture sensitive and hygroscopic. Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 368-39-8
Molecular Formula C6H15BF4O
Molecular Weight (g/mol) 189.99
MDL Number MFCD00044423
InChI Key IYDQMLLDOVRSJJ-UHFFFAOYSA-N
Synonym triethyloxonium tetrafluoroborate, triethyloxonium fluoroborate, triethyloxonium fluoborate, triethyloxonium borofluoride, triethoxonium fluoroborate, meerwein's reagent, triethyloxidanium tetrafluoroborate, unii-z0b19dd36j, triethyloxonium tetraflouroborate, boron tetrafluoride triethyl oxonium
PubChem CID 2723982
IUPAC Name triethyloxidanium;tetrafluoroborate
SMILES F[B-](F)(F)F.CC[O+](CC)CC

Specifications

Melting Point 84°C to 89°C
Assay Percent Range 95%, Stabilized with 3-5% diethyl ether
Linear Formula (CH3CH2)3OBF4
Quantity 100 g
UN Number UN2925
Beilstein 3598090
Sensitivity Moisture sensitive
Solubility Information Soluble in dichloromethane.
Formula Weight 189.99
Percent Purity 95%
Chemical Name or Material Triethyloxonium tetrafluoroborate, stabilized with 3 to 5% diethyl ether
Show More Show Less
Hazard Category H314-H335-H336
Hazard Statement GHS H Statement
H228-H314
Flammable solid.
Causes severe skin burns and eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLIDS, CORROSIVE, ORGANIC, N.O.S.
EINECSNumber 206-705-1
RTECSNumber RR4584700
TSCA Yes
storageNote1 Moisture sensitive
Recommended Storage Store at -20°C; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.