Learn More
Sigma Aldrich Fine Chemicals Biosciences Trifluoroacetic acid eluen
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 80457
Encompass
Trifluoroacetic acid is commonly used as a mobile phase additive in liquid chromatography-mass spectrometry (LC-MS). FeaturesExtensive QC testing ensures the highest specifications.AAAAAAAAOptimized to improve ionization and reSOLUTIONon.AAAAAAAA Extremely low levels of inorganic and organic impurities.AAAAAAAManufactured specifically for accurate and fast LC-MS.
Catalog No. 50-290-7588
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.