Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trifluoroacetic Acid, LC-MS Ultra, Solstice

Catalog No. 60044841 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 mL
2 mL
50 mL
Packaging:
Ampoule
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-044-841 50 mL Glass Bottle
60-048-600 1 mL Ampoule
60-048-601 2 mL Ampoule
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-044-841 Supplier Solstice Supplier No. 1426450ML

LC-MS Ultra, Eluent additive for UHPLC-MS

  • For the most demanding LC-MS and UHPLC applications
  • Tested for UHPLC-MS suitability
  • Highest solvent purity for low detection limits
  • Extremely low MS baseline noise
  • Exemplary lot-to-lot reproducibility
  • Low levels of trace metals
  • Packaged in clear borosilicate glass bottles
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76-05-1
Molecular Formula C2HF3O2
Molecular Weight (g/mol) 114.02
MDL Number MFCD00004169
InChI Key DTQVDTLACAAQTR-UHFFFAOYSA-N
Synonym trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova
PubChem CID 6422
ChEBI CHEBI:45892
SMILES OC(=O)C(F)(F)F

Specifications

Melting Point -15°C
Boiling Point 72°C
Color Colorless
Linear Formula CF3COOH
UN Number UN2699
Beilstein 742035
Solubility Information Soluble in water
IUPAC Name 2,2,2-trifluoroacetic acid
Formula Weight 114.02g/mol
Grade LC-MS Ultra
Density 1.489g/mL at 20°C
Quantity 50 mL
Flash Point >100°C (>212°F)
Packaging Glass Bottle
Physical Form Liquid
Refractive Index n20/D 1.3(lit.)
Vapor Density 3.9 (vs air)
Vapor Pressure 97.5mmHg (20°C)
Chemical Name or Material Trifluoroacetic Acid
Show More Show Less
Hazard Statement P273-P280-P305 + P351 + P338-P310
Precautionary Statement H314-H332-H412
DOTInformation 8/PGI
EINECSNumber 200-929-3
RTECSNumber AJ9625000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.