Learn More
Synquest Labs Inc Trifluoromethane, 75-46-7, MFCD00000446, 50g
Supplier: Synquest Labs Inc 110030350G
Molecular Formula: CHF3, Purity: 98%, Molecular Weight: 70.01, Melting Point: -160°C, Boiling Point: -84°C
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.