Learn More
Sigma Organic Chemistry Trifluoromethanesulfonic anhydride solution | 100ML | 358-23-6 | MFCD00000408
Supplier: Sigma Organic Chemistry 704083100ML
Trifluoromethanesulfonic anhydride solution, 100ML
About This Item:
Density: 1.359 g/mL at 25 C
Refractive Index: n20/D 1.402
Storage: 2-8C
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.