Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trifluoromethanesulfonic Acid 98.0+%, TCI America™
SDP

Catalog No. T0751250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
25 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0751250G 250 g
T075110G 10 g
T075125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T0751250G Supplier TCI America Supplier No. T0751250G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1493-13-6
Molecular Formula CHF3O3S
Molecular Weight (g/mol) 150.07
MDL Number MFCD00007514
InChI Key ITMCEJHCFYSIIV-UHFFFAOYSA-N
Synonym triflic acid, perfluoromethanesulfonic acid, trifluoromethanesulphonic acid, tfmsa, trifluoromethane sulfonic acid, tfoh, methanesulfonic acid, trifluoro, hotf, trifluoromethylsulfonic acid, trimsylate
PubChem CID 62406
ChEBI CHEBI:48511
IUPAC Name trifluoromethanesulfonic acid
SMILES OS(=O)(=O)C(F)(F)F

Specifications

Color Yellow
Boiling Point 162°C
Quantity 250 g
UN Number 3265
Formula Weight 150.07
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Trifluoromethanesulfonic Acid
EINECSNumber (2)-2809
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.