Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Trifluoromethanesulfonic Anhydride

Catalog No. 9173725ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
91-737-25ML 25 mL
91-737-100ML 100 mL
91-737-5ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 91-737-25ML Supplier Sigma Aldrich Supplier No. 9173725ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Trifluoromethanesulfonic anhydride can be used in the preparation of azido sugars such as 6-azido fucose. It can also be used as a promoter for the Friedel-Crafts acylation of aromatic compounds with carboxylic acids in the absence of catalyst.Tf2O in combination with:Diphenyl sulfoxide, forms an efficient promoter system for the activation of thioglycosides for glycosylation reactions.2-chloropyridine, can be used for secondary amide activation.Potassium iodide, can effectively mediate the deoxygenation of sulfoxides to form sulfides.Hydrogen peroxide, for the selective oxidation of sulfanes to sulf-íoxides in the presence of oxidatively sensitive functional groups.Tf2O can react with:Ketones in the presence of a base to form vinyl triflates.Magnesium alkoxides to form corresponding symmetrical or unsymmetrical ethers.Sodium azide for the in situ preparation of triflyl azide.Electrophilic trifyl source. Reviews

Specifications

CAS 358-23-6
Density 1.677 g/mL at 25°C
Boiling Point 81°C to 83°C
Molecular Formula C2F6O5S2
Refractive Index n20/D 1.321
Linear Formula (CF3SO2)2O
MDL Number MFCD00000408
Quantity 25 mL
Synonym Tf2O; Triflic anhydride
Molecular Weight (g/mol) 282.14 g/mol
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.