Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trifluoromethanesulfonic anhydride, 98+%

Catalog No. AC175060010
Change view
Click to view available options
Quantity:
1 mL
50 mL
250 mL
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC175060010 1 mL Glass bottle
AC175060500 50 mL Glass bottle
AC175062500 250 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC175060010 Supplier Thermo Scientific Chemicals Supplier No. 175060010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

Trifluoromethanesulfonic anhydride, 98+%, Quantity: 250mL, Packaging: Glass bottle, Boiling Point: 80.0 deg.C, Colorless to Brown, Molecular Weight: 282.13, Percent Purity: 98+%, Assay Percent Range: 98+%, Beilstein: 03, IV, 35, CAS: 358-23-6, ChEBI: CHEBI:48509, Density: 1.7000g/mL | CAS: 358-23-6 | C2F6O5S2 | 282.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent, Coupling reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 358-23-6
MDL Number MFCD00000408
InChI Key WJKHJLXJJJATHN-UHFFFAOYSA-N
Synonym trifluoromethanesulfonic anhydride, triflic anhydride, trifluoromethanesulphonic anhydride, trifluoromethanesulfonic acid anhydride, methanesulfonic acid, trifluoro-, anhydride, trifluoromethanesulphonic acid anhydride, unii-8w034lhg1u, tf2o, trifluoromethane sulfonyl trifluoromethanesulfonate, trifluoromethane sulfonic anhydride
PubChem CID 67749
ChEBI CHEBI:48509
IUPAC Name trifluoromethylsulfonyl trifluoromethanesulfonate
SMILES C(F)(F)(F)S(=O)(=O)OS(=O)(=O)C(F)(F)F

Specifications

Color Colorless to Brown
Density 1.7000g/mL
Boiling Point 80.0°C
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Molecular Formula C2F6O5S2
Refractive Index 1.3200 to 1.3230
Linear Formula (CF3SO2)2O
Quantity 1 mL
Beilstein 03, IV, 35
Fieser 04,533; 05,702; 06,618; 07,390; 10,419; 11,560; 12,532; 14,324; 15,339; 16,357; 17,156
Specific Gravity 1.7
Merck Index 15, 9850
Solubility Information Solubility in water: reacts viontly with water
Molecular Weight (g/mol) 282.13
Formula Weight 282.13
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material Trifluoromethanesulfonic anhydride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Reacts violently with water.
May be corrosive to metals.
Precautionary Statement GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove
EINECSNumber 206-616-8

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.