Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trifluoromethanesulfonic Anhydride 98.0+%, TCI America™
SDP

Catalog No. T1100250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
25 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1100250G 250 g
T110010G 10 g
T110025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T1100250G Supplier TCI America Supplier No. T1100250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 358-23-6
Molecular Formula C2F6O5S2
Molecular Weight (g/mol) 282.127
MDL Number MFCD00000408
InChI Key WJKHJLXJJJATHN-UHFFFAOYSA-N
Synonym trifluoromethanesulfonic anhydride, triflic anhydride, trifluoromethanesulphonic anhydride, trifluoromethanesulfonic acid anhydride, methanesulfonic acid, trifluoro-, anhydride, trifluoromethanesulphonic acid anhydride, unii-8w034lhg1u, tf2o, trifluoromethane sulfonyl trifluoromethanesulfonate, trifluoromethane sulfonic anhydride
PubChem CID 67749
ChEBI CHEBI:48509
IUPAC Name trifluoromethylsulfonyl trifluoromethanesulfonate
SMILES C(F)(F)(F)S(=O)(=O)OS(=O)(=O)C(F)(F)F

Specifications

Melting Point -80°C
Color Colorless
Boiling Point 84°C
Quantity 250 g
UN Number 3265
Formula Weight 282.13
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Trifluoromethanesulfonic Anhydride
EINECSNumber (2)-3807
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.