Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beckman Coulter™ Triglyceride Reagents

Catalog No. 01801035 Shop All Beckman Coulter Products
Change view
Click to view available options
Includes:
R1 4 x 167 mL, R2 4 x 43 mL
R1 4 x 50 mL, R2 4 x 12.5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
01-801-035 R1 4 x 50 mL, R2 4 x 12.5 mL
01-801-075 R1 4 x 167 mL, R2 4 x 43 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 01-801-035 Supplier Beckman Coulter™ Supplier No. OSR61118
Add to Cart
Add to Cart
This item is not returnable. View return policy

Triglyceride Reagents for use with Beckman Coulter analyzers.

  • For use with Beckman Coulter analyzers
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Beckman Coulter Analyzers
Includes R1 4 x 50 mL, R2 4 x 12.5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.