Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triisopropylsilyl Chloride 95.0+%, TCI America™
SDP

Catalog No. T1078250ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
250 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1078250ML 250 mL
T10785ML 5 mL
T107825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T1078250ML Supplier TCI America Supplier No. T1078250ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13154-24-0
Molecular Formula C9H21ClSi
Molecular Weight (g/mol) 192.80
MDL Number MFCD00009656
InChI Key KQIADDMXRMTWHZ-UHFFFAOYSA-N
Synonym triisopropylsilyl chloride, chlorotriisopropylsilane, triisopropylchlorosilane, chlorotris propan-2-yl silane, chloro triisopropyl silane, silane, chlorotris 1-methylethyl, tipscl, triisopropylsilylchloride, triisopropyl chlorosilane, triisopropylchlorosilane, redistilled
PubChem CID 139400
IUPAC Name chlorotris(propan-2-yl)silane
SMILES CC(C)[Si](Cl)(C(C)C)C(C)C

Specifications

Color Yellow
Boiling Point 96°C
Quantity 250 mL
UN Number 2987
Formula Weight 192.80
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Triisopropylsilyl Chloride
EINECSNumber (2)-2041
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.