Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Trilostane

Catalog No. AC464380010
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC464380010 1 g
AC464382500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC464380010 Supplier Thermo Scientific Chemicals Supplier No. 464380010
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13647-35-3
Molecular Formula C20H27NO3
Molecular Weight (g/mol) 329.44
InChI Key KVJXBPDAXMEYOA-CXANFOAXSA-N
IUPAC Name (1S,2R,6R,8S,11S,12S,15S,16S)-5,15-dihydroxy-2,16-dimethyl-7-oxapentacyclo[9.7.0.0²,⁸.0⁶,⁸.0¹²,¹⁶]octadec-4-ene-4-carbonitrile
SMILES C[C@]12CC[C@H]3[C@@H](CC[C@@]45O[C@@H]4C(O)=C(C[C@]35C)C#N)[C@@H]1CC[C@@H]2O

Specifications

Melting Point 284.0°C
Quantity 1 g
Formula Weight 329.43
Chemical Name or Material Trilostane
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Causes skin irritation.

Causes serious eye irritation.

Suspected of damaging fertility or the unborn child.

Precautionary Statement GHS P Statement IF ON SKIN: Wash with plenty of soap and water.

If skin irritation occurs: Get medical advice/attention.

Obtain special instructions before use.

Wear protective gloves/protective clothing/eye protection/face protection.

IF exposed or concerned: Get medical advice/attention.

EINECSNumber 237-133-0

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.