Learn More
Invitrogen™ TRIM Monoclonal Antibody (TRIM-04), PE

Description
This antibody recognizes an intracellular epitope of T cell receptor-interacting molecule (TRIM), a 30 kDa adaptor protein expressed by T cells. This antibody will not cross-react with mouse.
Specifications
Specifications
| Antigen | TRIM |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | TRIM-04 |
| Concentration | 0.1 mg/mL |
| Conjugate | PE |
| Formulation | PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 |
| Gene | TRAT1 |
| Gene Accession No. | Q6PIZ9 |
| Gene Alias | C030046M14Rik; HSPC062; pp29/30; T cell receptor associated transmembrane adaptor 1; T cell receptor interacting molecule; T-cell receptor interacting molecule; T-cell receptor-associated transmembrane adapter 1; T-cell receptor-interacting molecule; TCRIM; Trat1; TRIM |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.