Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIM, R&D Systems™
SDP

Catalog No. 091950 Shop All Novus Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-195-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 09-195-0 Supplier R&D Systems Supplier No. 0919/50

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. nNOS/iNOS inhibitor

A potent inhibitor of neuronal and inducible NO synthases, with much lower affinity for the endothelial isoform (displays IC50 values of 28.2, 27.0 and 1057.5 μM respectively). Antinociceptive in vivo.

Chemical Identifiers

CAS 25371-96-4
Molecular Formula C10H7F3N2
Molecular Weight (g/mol) 212.175
InChI Key WZBWBNCQUTXYEL-UHFFFAOYSA-N
Synonym trim, 1-2-trifluoromethyl phenyl imidazole, 1-2-trifluoromethylphenyl imidazole, 1-2-trifluoromethyl phenyl-1h-imidazole, 1h-imidazole,1-2-trifluoromethyl phenyl, dsstox_cid_25217, dsstox_rid_80756, dsstox_gsid_45217, 2-trifluoromethyl phenyl imidazole
PubChem CID 1359
IUPAC Name 1-[2-(trifluoromethyl)phenyl]imidazole
SMILES C1=CC=C(C(=C1)C(F)(F)F)N2C=CN=C2

Specifications

Quantity 50 mg
Formula Weight 212.17
Percent Purity >99%
Product Line Tocris
Chemical Name or Material TRIM
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.