Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRIM10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531291
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531291 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531291 Supplier Invitrogen™ Supplier No. PA531291
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: NT2D1, IMR32, U87-MG, MCF-7. Predicted reactivity: Rhesus Monkey (95%), Chimpanzee (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. This protein localizes to cytoplasmic bodies. Studies in mice suggest that this protein plays a role in terminal differentiation of erythroid cells. Alternate splicing of this gene generates two transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIM10
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.98 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene TRIM10
Gene Accession No. Q9UDY6
Gene Alias AI324236; B30-RING finger protein; BB139825; Hematopoietic RING finger 1; Herf1; RFB30; RING finger protein 9; Rnf9; TRIM10; tripartite motif containing 10; tripartite motif protein 10; tripartite motif-containing 10; tripartite motif-containing protein 10; Zn-finger protein
Gene Symbols TRIM10
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human TRIM10. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10107
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.