Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRIM15 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540946
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540946 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540946 Supplier Invitrogen™ Supplier No. PA540946
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: RDEIEDVKCQ EDQKLQVLLT QIESKKHQVE TAFERLQQEL EQQRCLLLAR Sequence homology: Cow: 86%; Horse: 86%; Human: 100%; Mouse: 93%; Rabbit: 79%; Rat: 86%.

The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to the cytoplasm. Alternatively spliced transcript variants have been described, but their biological validity has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIM15
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TRIM15
Gene Accession No. Q9C019
Gene Alias 1810012B10Rik; LOC100910806; RING finger protein 93; RNF93; TRIM15; tripartite motif containing 15; tripartite motif protein 15; tripartite motif-containing 15; tripartite motif-containing protein 15; tripartite motif-containing protein 15-like; zinc finger protein 178; Zinc finger protein B7; ZNF178; ZNFB7
Gene Symbols TRIM15
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human TRIM15 (aa 141-190).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 89870
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.