Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRIM2 Monoclonal Antibody (OTI3B5)
GREENER_CHOICE

Catalog No. PIMA525207
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIMA525207 100 μL
1 options

Catalog No. PIMA525207

Supplier: Invitrogen™ MA525207

Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TRIM2 Monoclonal Antibody for Western Blot, ICC/IF

The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to cytoplasmic filaments. Its function has not been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIM2
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI3B5
Concentration 0.66 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Trim2
Gene Accession No. Q9C040
Gene Alias CMT2R; E3 ubiquitin-protein ligase TRIM2; KIAA0517; mKIAA0517; narf; Neural activity-related RING finger protein; RING finger protein 86; RING-type E3 ubiquitin transferase TRIM2; RNF86; TRIM2; tripartite motif containing 2; tripartite motif protein 2; tripartite motif protein TRIM2; tripartite motif-containing 2; tripartite motif-containing protein 2
Gene Symbols Trim2
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-100 and 645-744 of TRIM2 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23321
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.