Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIM21/SSA1 Antibody, DyLight 350, R&D Systems™
SDP

Catalog No. NB262841
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB262841 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB262841 Supplier R&D Systems Supplier No. NBP137009UV

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

TRIM21/SSA1 Polyclonal specifically detects TRIM21/SSA1 in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative).

Specifications

Antigen TRIM21/SSA1
Applications Immunoprecipitation, Western Blot (Negative)
Classification Polyclonal
Conjugate DyLight 350
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias 52 kDa ribonucleoprotein autoantigen Ro/SS-A, 52 kDa Ro/SSA, E3 ubiquitin-protein ligase TRIM21, EC 6.3.2.-, rhTRIM21, RNF81RING finger protein 81, ro(SS-A), RO52SS-A, Sicca syndrome antigen A, Sjoegren syndrome type A antigen, Sjogren syndrome antigen A1 (52kDa, ribonucleoprotein autoantigen SS-A/Ro), SSA1SSA, tripartite motif containing 21,52 kDa Ro protein, tripartite motif-containing 21, Tripartite motif-containing protein 21
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 250 and 300 of human Sjogren syndrome antigen A1 using the numbering given in entry NP_003132.2 (GeneID 6737).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 6737
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.