Learn More
Invitrogen™ TRIM29 Polyclonal Antibody

Description
Predicted to react with Mouse, Rat, Chimpanzee and Canine.
Specifications
Specifications
| Antigen | TRIM29 |
| Applications | ELISA, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.2% gelatin and 0.05% sodium azide |
| Gene | Trim29 |
| Gene Accession No. | Q14134 |
| Gene Alias | 1110047J21Rik; 2810431N19Rik; 4732461M22Rik; AI119726; Ataxia telangiectasia group D-associated protein; ataxia-telangiectasia group D-associated protein; ATDC; EGK_06995; FLJ36085; Trim29; tripartite motif containing 29; tripartite motif protein 29; tripartite motif protein TRIM29; tripartite motif-containing 29; tripartite motif-containing protein 29 |
| Gene Symbols | Trim29 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.