Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIM33 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501736126 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6126 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6126 Supplier Proteintech Group Inc Supplier No. 553741AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is specific to TRIM33. The calculated molecular weight of TRIM33 is 122 kDa, but the molecular weight of detected is about 140-150 kDa as the same as the result from the paper.

TIF1 gamma is a member of the Transcriptional Intermediary Factor 1 family, which serve as coactivators and corepressors. Its structure is similar to TIF1 alpha and TIF1 beta, exhibiting multiple domains (RING finger, coiled coil, B boxes, PHD/TTC, and bromodomain). TIF1 gamma functions in cell differentiation and development, playing a role in differentiation of hematopoietic cells.

Specifications

Antigen TRIM33
Applications Western Blot
Classification Polyclonal
Concentration 0.43 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene TRIM33
Gene Accession No. Q9UPN9
Gene Alias Ectodermin homolog, KIAA1113, Protein Rfg7, PTC7, RET fused gene 7 protein, RFG7, TF1G, TIF1 gamma, TIF1G, TIF1GAMMA, TIFGAMMA, TRIM33, tripartite motif containing 33
Gene Symbols TRIM33
Host Species Rabbit
Immunogen Peptide
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51592
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.