Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TRIM38 Monoclonal Antibody (OTI16D9)

Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA526236
Description
TRIM38 Monoclonal Antibody for Western Blot, IHC (P)
This gene encodes a member of the tripartite motif (TRIM) family. The encoded protein contains a RING-type zinc finger, B box-type zinc finger and SPRY domain. The function of this protein has not been identified. A pseudogene of this gene is located on the long arm of chromosome 4.
Specifications
| TRIM38 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O00635 | |
| TRIM38 | |
| Human recombinant protein fragment corresponding to amino acids 1-265 of TRIM38 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human, Monkey | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI16D9 | |
| Unconjugated | |
| TRIM38 | |
| E3 ubiquitin-protein ligase TRIM38; Gm23; hypothetical protein LOC531022; MGC8946; RING finger protein 15; RING-type E3 ubiquitin transferase TRIM38; RNF15; Ro/SSA ribonucleoprotein homolog; RORET; TRIM38; tripartite motif containing 38; tripartite motif-containing 38; tripartite motif-containing protein 38; tripartite motif-containing protein 38; E3 ubiquitin-protein ligase TRIM38; tripartite motif-containing protein 38; LOW QUALITY PROTEIN: E3 ubiquitin-protein ligase TRIM38; zinc finger protein RoRet | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 10475 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction