Learn More
TRIM38 Mouse anti-Human, Clone: OTI113D3, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF505981

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a member of the tripartite motif (TRIM) family. The encoded protein contains a RING-type zinc finger, B box-type zinc finger and SPRY domain. The function of this protein has not been identified. A pseudogene of this gene is located on the long arm of chromosome 4.Specifications
TRIM38 | |
Monoclonal | |
Unconjugated | |
TRIM38 | |
RNF15, RORET | |
Mouse | |
Affinity Chromatography | |
RUO | |
10475 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI113D3 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
O00635 | |
TRIM38 | |
Human recombit protein fragment corresponding to amino acids 1-265 of human TRIM38 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.