Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIM45 Mouse anti-Human, Clone: OTI2B11, lyophilized, TrueMAB™
SDP

Catalog No. 501664018
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-166-4018 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-166-4018 Supplier OriGene Supplier No. CF505917
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO

TRIM45 is a member of the tripartite motif family. It may function as a transcriptional repressor of the mitogen-activated protein kinase pathway. Alternatively spliced transcript variants have been described. TRIM45 belongs to a family of tripartite motif (TRIM) proteins that play important roles in a variety of cellular functions, including cell proliferation, differentiation, development, oncogenesis, and apoptosis.

Specifications

Antigen TRIM45
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI2B11
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene TRIM45
Gene Accession No. Q9H8W5
Gene Alias RNF99
Gene Symbols TRIM45
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 288-580 of human TRIM45 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80263
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.