Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIM56 Mouse anti-Human, Clone: OTI8E12, liquid, TrueMAB™
SDP

Catalog No. 501682475
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-2475 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-2475 Supplier OriGene Supplier No. TA808934
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

E3 ubiquitin-protein ligase that plays a key role in innate antiviral immunity. In response to pathogen- and host-derived double-stranded DNA (dsDNA), targets TMEM173/STING to ′Lys-63′-linked ubiquitination, thereby promoting its homodimerization, a step required for the production of type I interferon IFN-beta Independently of its E3 ubiquitin ligase activity, positive regulator of TLR3 signaling. Potentiates extracellular double stranded RNA (dsRNA)-induced expression of IFNB1 and interferon-stimulated genes ISG15, IFIT1/ISG56, CXCL10, OASL and CCL5/RANTES. Promotes establishment of an antiviral state by TLR3 ligand and TLR3-mediated chemokine induction following infection by hepatitis C virus.

Specifications

Antigen TRIM56
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI8E12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene TRIM56
Gene Accession No. Q9BRZ2
Gene Alias RNF109
Gene Symbols TRIM56
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 311-554 of human TRIM56 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81844
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.