Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRIM59 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542367
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542367 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542367 Supplier Invitrogen™ Supplier No. PA542367
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: LEKVDDVRQH VQILKQRPLP EVQPVEIYPR VSKILKEEWS RTEIGQIKNV Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 93%; Horse: 86%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 86%; Rat: 100%.

May serve as a multifunctional regulator for innate immune signaling pathways. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIM59
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TRIM59
Gene Accession No. Q8IWR1
Gene Alias 2310035M22Rik; 2700022F13Rik; IFT80L; MRF1; RGD1311956; RING finger 1; RING finger protein 1; RING finger protein 104; RNF104; TRIM57; TRIM59; tripartite motif containing 59; tripartite motif-containing 57; tripartite motif-containing 59; tripartite motif-containing protein 59; tripartite motif-containing protein 59-like protein; TSBF1; tumor suppressor TSBF1; Tumor suppressor TSBF-1; tumor suppressor TSBF1 homolog; Unknown (protein for MGC:137058)
Gene Symbols TRIM59
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human TRIM59.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 286827
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.