Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethoxy(phenylethyl)silane (mixture of 1-phenylethyl- and 2-phenylethyl-) 95.0+%, TCI America™
SDP

Catalog No. T3542100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3542100G 100 g
T354225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T3542100G Supplier TCI America Supplier No. T3542100G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 49539-88-0
Molecular Formula C11H18O3Si
Molecular Weight (g/mol) 226.347
MDL Number MFCD00053852
InChI Key UBMUZYGBAGFCDF-UHFFFAOYSA-N
PubChem CID 170789
IUPAC Name trimethoxy(2-phenylethyl)silane
SMILES CO[Si](CCC1=CC=CC=C1)(OC)OC

Specifications

Boiling Point 122°C
Quantity 100 g
Formula Weight 226.35
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Trimethoxy(phenylethyl)silane (mixture of 1-phenylethyl- and 2-phenylethyl-)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.